accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONHNH2HCl

CAS RN 59146-68-8

IUPAC Name: [3-(benzyloxy)phenyl]hydrazine hydrochloride
Synonyms: [3-(benzyloxy)phenyl]hydrazine hydrochloride
Molecular Weight (g/mol): 250.73
Molecular Formula: C13H15ClN2O
InChi Key: MMMGCMOISVZVKZ-UHFFFAOYSA-N
SMILES: Cl.NNC1=CC(OCC2=CC=CC=C2)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results