accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COClO

CAS RN 592-34-7

IUPAC Name: butyl carbonochloridate
Synonyms: carbonochloridic acid, butyl ester
Molecular Weight (g/mol): 136.58
Molecular Formula: C5H9ClO2
InChi Key: NRDQFWXVTPZZAZ-UHFFFAOYSA-N
SMILES: CCCCOC(Cl)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)