accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH2

CAS RN 592-41-6

IUPAC Name: hex-1-ene
Synonyms: 1-hexene
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: LIKMAJRDDDTEIG-UHFFFAOYSA-N
SMILES: CCCCC=C
Molecular Weight (g/mol) 
  • (6)
  • (3)
  • (5)
Quantity 
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (3)
  • (6)
Percent Purity 
  • (4)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (3)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (4)
  • (3)
  • (2)
  • (7)
  • (1)
  • (1)