accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 592-82-5

IUPAC Name: 1-isothiocyanatobutane
Synonyms: butyl isothiocyanate
Molecular Weight (g/mol): 115.19
Molecular Formula: C5H9NS
InChi Key: LIMQQADUEULBSO-UHFFFAOYSA-N
SMILES: CCCCN=C=S
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results