accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 592-85-8

HgSSNN

CAS RN 592-85-8

IUPAC Name: mercury(2+) dipotassium tetrakis(cyanosulfanide)
Synonyms: mercury(II) thiocyanate
Molecular Weight (g/mol): 511.1
Molecular Formula: C4HgK2N4S4
InChi Key: HWXOCCNUUYRBKP-UHFFFAOYSA-J
SMILES: [K+].[K+].[Hg++].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)