CAS RN 592-88-1
IUPAC Name:
3-(prop-2-en-1-ylsulfanyl)prop-1-ene
Synonyms:
oil garlic allium sativum oil oil of garlic (substance) thioallyl ether garlic oil oil of garlic garlic oil preparation
Molecular Weight (g/mol):
114.21
Molecular Formula:
C6H10S
InChi Key:
UBJVUCKUDDKUJF-UHFFFAOYSA-N
SMILES:
C=CCSCC=C