accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2ClN

CAS RN 5922-60-1

IUPAC Name: 2-amino-5-chlorobenzonitrile
Synonyms: 2-amino-5-chlorobenzonitrile
Molecular Weight (g/mol): 152.58
Molecular Formula: C7H5ClN2
InChi Key: QYRDWARBHMCOAG-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C=C1C#N
Quantity 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)