accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH2SnH3CCH3OHH3COSnH3CCH3OHCH3O

CAS RN 5926-79-4

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 435.68
Molecular Formula: C8H22O5Sn2
InChi Key: PTOQDQIJSRHODZ-UHFFFAOYSA-N
SMILES: O.C[SnH2]C.C[SnH2]C.CC(O)=O.CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results