accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SOH3C

CAS RN 5928-48-3

IUPAC Name: 4,5-dimethylthiophene-2-carbaldehyde
Synonyms: 4,5-dimethylthiophene-2-carbaldehyde
Molecular Weight (g/mol): 140.2
Molecular Formula: C7H8OS
InChi Key: QSBBXKVGLJSGAJ-UHFFFAOYSA-N
SMILES: CC1=C(C)C=C(S1)C=O
Boiling Point 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)