accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 593-66-8

IUPAC Name: iodoethene
Synonyms: vinyl iodide
Molecular Weight (g/mol): 153.95
Molecular Formula: C2H3I
InChi Key: GHXZPUGJZVBLGC-UHFFFAOYSA-N
SMILES: IC=C
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)