CAS RN 593-84-0
Guanidine Thiocyanate (White Crystalline Powder), Fisher BioReagents™
CAS: 593-84-0 Molecular Formula: C2H6N4S Synonym: Guanidinium Isothiocyanate
Thermo Scientific Chemicals Guanidine thiocyanate, 99%, for biochemistry
CAS: 593-84-0 Molecular Formula: CH5N3·CHNS Molecular Weight (g/mol): 118.16 MDL Number: MFCD00013027
Thermo Scientific Chemicals Guanidine thiocyanate, Molecular Biology Grade
CAS: 593-84-0 Molecular Formula: CH5N3·CHNS MDL Number: MFCD00013027 Synonym: Guanidine rhodanide
MP Biomedicals™ Geneclean™ Spin Kit 200 bp to 300 kb
Used to purify fragments of DNA from 200 bp to 300 kbp from solutions and agarose gels. The GENECLEAN™ SPIN kit uses SPIN filters, which prevent silica particle carry-over into cleaned DNA. Glassmilk™ slurry allows flexibility of reaction size and volume.
Macherey-Nagel™ Lysis Buffer LBP
Additional buffer solutions for use with NucleoSpin™ RNA Plus Kits, for efficient and quick RNA isolation from cells and tissues.
Macherey-Nagel™ Lysis Buffer DL
Lysis Buffer DL to be used with NucleoSpin RNA Blood kits
Macherey-Nagel™ Binding Buffer NT
Binding Buffer NT to be used with NucleoSpin 8 / 96 PCR Clean‐up kits
Guanidine Thiocyanate, TRC
CAS: 593-84-0 Molecular Formula: C H5 N3 . C H N S Molecular Weight (g/mol): 118.16 Synonym: Guanidine Thiocyanate,Thiocyanic acid, compd. with guanidine (1:1),Guanidine thiocyanate (6CI),Guanidine, monothiocyanate (8CI,9CI),Buffer AVL,Buffer ML,Buffer MTL,Buffer RLT,GTC,Guanidine isothiocyanate,Guanidinium isothiocyanate,Guanidinium thiocyanate,NSC 2119,RLT IUPAC Name: guanidine;thiocyanic acid SMILES: NC(=N)N.SC#N
Macherey-Nagel™ Wash Buffer MR3
Additional buffer solutions (e.g., lysis buffer MR1, wash buffer MR3) for use in magnetic bead-based isolation of RNA from cell and tissue samples using the NucleoMag™ RNA kit.
Macherey-Nagel™ Lysis Buffer ML
Lysis Buffer ML to be used with NucleoSpin miRNA kits.
Macherey-Nagel™ Buffers for NucleoSpin™ Virus Kits
Supplemental buffer solutions (e.g., wash and lysis buffers) for use with NucleoSpin™ Virus kits for the rapid isolation of viral nucleic acids (RNA and DNA).
Macherey-Nagel™ Binding Buffer NTI
Binding Buffer NTI to be used with NucleoSpin Gel and PCR Clean‐up or NucleoSpin Gel and PCR Clean‐up Midi / Maxi kits.
MP Biomedicals™ Geneclean™ Glassmilk™ Silica Matrix
Geneclean™ Glassmilk™ Silica Matrix allows flexibility of reaction size and volume. Glassmilk™ slurry is a specially prepared aqeuous suspension of propietary silica matrix.
- 1
- 2