accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 59377-19-4

OCNO

CAS RN 59377-19-4

IUPAC Name: 1-isocyanato-4-phenoxybenzene
Synonyms: 1-isocyanato-4-phenoxybenzene
Molecular Weight (g/mol): 211.22
Molecular Formula: C13H9NO2
InChi Key: PNBUGOFIKAHZRW-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=C(OC2=CC=CC=C2)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results