accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SnCH3CH3H3C

CAS RN 594-27-4

IUPAC Name: tetramethylstannane
Synonyms: tetramethyltin tetramethyl stannane SnMe4
Molecular Weight (g/mol): 178.85
Molecular Formula: C4H12Sn
InChi Key: VXKWYPOMXBVZSJ-UHFFFAOYSA-N
SMILES: C[Sn](C)(C)C
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)