accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 594-34-3

IUPAC Name: 1,2-dibromo-2-methylpropane
Synonyms: 1,2-dibromo-2-methylpropane
Molecular Weight (g/mol): 215.92
Molecular Formula: C4H8Br2
InChi Key: SDTXSEXYPROZSZ-UHFFFAOYSA-N
SMILES: CC(C)(Br)CBr
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results