accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClSClClCl

CAS RN 594-42-3

IUPAC Name: trichloro(chlorosulfanyl)methane
Synonyms: perchloromethyl mercaptan
Molecular Weight (g/mol): 185.87
Molecular Formula: CCl4S
InChi Key: RYFZYYUIAZYQLC-UHFFFAOYSA-N
SMILES: ClSC(Cl)(Cl)Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)