accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 595-90-4

Sn

CAS RN 595-90-4

IUPAC Name: tetraphenylstannane
Synonyms: tetraphenyl tin
Molecular Weight (g/mol): 427.13
Molecular Formula: C24H20Sn
InChi Key: CRHIAMBJMSSNNM-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[Sn](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)