accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 59528-27-7

NH2IHCl

CAS RN 59528-27-7

IUPAC Name: 1-(4-iodophenyl)methanamine hydrochloride
Synonyms: 1-(4-iodophenyl)methanamine hydrochloride
Molecular Weight (g/mol): 269.51
Molecular Formula: C7H9ClIN
InChi Key: GBJMURRFWZREHE-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results