accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FHN

CAS RN 59611-52-8

IUPAC Name: 6-fluoro-1,2,3,4-tetrahydroquinoline
Synonyms: 6-fluoro-1,2,3,4-tetrahydroquinoline
Molecular Weight (g/mol): 151.18
Molecular Formula: C9H10FN
InChi Key: NECDDBBJVCNJNS-UHFFFAOYSA-N
SMILES: FC1=CC2=C(NCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results