accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5965-38-8

IUPAC Name: (5Z)-5-[(3-chloro-4-hydroxy-5-methoxyphenyl)methylidene]-1-(prop-2-en-1-yl)-2-sulfanylidene-1,3-diazinane-4,6-dione
Synonyms: (5Z)-5-[(3-chloro-4-hydroxy-5-methoxyphenyl)methylidene]-1-(prop-2-en-1-yl)-2-sulfanylidene-1,3-diazinane-4,6-dione
Molecular Weight (g/mol): 352.79
Molecular Formula: C15H13ClN2O4S
InChi Key: YOKXELRIGMPSIG-UITAMQMPSA-N
SMILES: COC1=CC(\C=C2\C(=O)NC(=S)N(CC=C)C2=O)=CC(Cl)=C1O
Grade 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results