accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 59652-95-8

HNOOOHBr(E/Z)

CAS RN 59652-95-8

IUPAC Name: 3-[(2-bromophenyl)carbamoyl]prop-2-enoic acid
Synonyms: 3-[(2-bromophenyl)carbamoyl]prop-2-enoic acid
Molecular Weight (g/mol): 270.08
Molecular Formula: C10H8BrNO3
InChi Key: CJHBOIXAXNGQNF-UHFFFAOYSA-N
SMILES: OC(=O)C=CC(=O)NC1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results