accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OZnCH3OOCH3OO

CAS RN 5970-45-6

IUPAC Name: zinc(2+) diacetate dihydrate
Synonyms: zinc(2+) diacetate dihydrate
Molecular Weight (g/mol): 219.5
Molecular Formula: C4H10O6Zn
InChi Key: BEAZKUGSCHFXIQ-UHFFFAOYSA-L
SMILES: O.O.[Zn++].CC([O-])=O.CC([O-])=O
Molecular Weight (g/mol) 
  • (22)
  • (9)
  • (3)
  • (3)
Quantity 
  • (1)
  • (8)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (4)
  • (1)
  • (3)
  • (8)
Percent Purity 
  • (5)
  • (4)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (6)
  • (7)
  • (1)
  • (4)
Grade 
  • (4)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (5)
  • (3)
  • (1)
  • (9)
  • (8)
  • (3)
  • (1)
  • (22)
  • (9)
  • (3)
  • (3)
  • (1)
  • (8)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (4)
  • (1)
  • (3)
  • (8)
  • (5)
  • (4)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (6)
  • (7)
  • (1)
  • (4)
  • (4)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)