accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOH

CAS RN 59748-18-4

IUPAC Name: 4'-(octyloxy)-[1,1'-biphenyl]-4-carboxylic acid
Synonyms: 4'-(octyloxy)-[1,1'-biphenyl]-4-carboxylic acid
Molecular Weight (g/mol): 326.44
Molecular Formula: C21H26O3
InChi Key: YNBBQLUKHHSKPW-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results