accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 598-21-0

IUPAC Name: 2-bromoacetyl bromide
Synonyms: acetyl bromide, bromo-
Molecular Weight (g/mol): 201.85
Molecular Formula: C2H2Br2O
InChi Key: LSTRKXWIZZZYAS-UHFFFAOYSA-N
SMILES: BrCC(Br)=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)