accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNHNH2O

CAS RN 598-50-5

IUPAC Name: methylurea
Synonyms: methylurea
Molecular Weight (g/mol): 74.08
Molecular Formula: C2H6N2O
InChi Key: XGEGHDBEHXKFPX-UHFFFAOYSA-N
SMILES: CNC(N)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)