accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNHNH2S

CAS RN 598-52-7

IUPAC Name: methylthiourea
Synonyms: N-methylthiourea
Molecular Weight (g/mol): 90.14
Molecular Formula: C2H6N2S
InChi Key: KQJQICVXLJTWQD-UHFFFAOYSA-N
SMILES: CNC(N)=S
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)