accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 598-62-9

MnOOO

CAS RN 598-62-9

IUPAC Name: manganese(2+) carbonate
Synonyms: manganese carbonate manganese(II) carbonate
Molecular Weight (g/mol): 114.95
Molecular Formula: CMnO3
InChi Key: XMWCXZJXESXBBY-UHFFFAOYSA-L
SMILES: [Mn++].[O-]C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)