accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CBrOHO

CAS RN 598-72-1

IUPAC Name: 2-bromopropanoic acid
Synonyms: α-bromopropionic acid
Molecular Weight (g/mol): 152.98
Molecular Formula: C3H5BrO2
InChi Key: MONMFXREYOKQTI-UHFFFAOYNA-N
SMILES: CC(Br)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)