accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 598-78-7

IUPAC Name: 2-chloropropanoic acid
Synonyms: 2-chloropropionic acid
Molecular Weight (g/mol): 108.52
Molecular Formula: C3H5ClO2
InChi Key: GAWAYYRQGQZKCR-UHFFFAOYNA-N
SMILES: CC(Cl)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)