accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5980-23-4

H2NOClCl

CAS RN 5980-23-4

IUPAC Name: 3,5-dichlorobenzamide
Synonyms: 3,5-dichlorobenzamide
Molecular Weight (g/mol): 190.02
Molecular Formula: C7H5Cl2NO
InChi Key: DELNZTRPJTUOIP-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(Cl)=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results