accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5980-97-2

IUPAC Name: (2,4,6-trimethylphenyl)boronic acid
Synonyms: 2,4,6-trimethylphenylboronic acid
Molecular Weight (g/mol): 164.01
Molecular Formula: C9H13BO2
InChi Key: BZXQRXJJJUZZAJ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(B(O)O)C(C)=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results