accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONH(R)(R)O(R)HO(R)HOO(R)O(R)HO(R)HO(S)HO(R)OHONOO

CAS RN 59837-14-8

IUPAC Name: N-[(2R,3R,5R,6R)-5-hydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)-4-{[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-3-yl]acetamide
Synonyms: N-[(2R,3R,5R,6R)-5-hydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)-4-{[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-3-yl]acetamide
Molecular Weight (g/mol): 504.45
Molecular Formula: C20H28N2O13
InChi Key: INMOOBMAIAWVBW-SLMMFOMASA-N
SMILES: CC(=O)N[C@H]1[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)O[C@H](CO)[C@H](O)C1O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results