CAS RN 5985-28-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac-Synephrine Hydrochloride, TRC
CAS: 5985-28-4 Molecular Formula: C9 H13 N O2 . Cl H Molecular Weight (g/mol): 203.67 Synonym: Benzenemethanol, 4-hydroxy-α-[(methylamino)methyl]-, hydrochloride (1:1),Benzenemethanol, 4-hydroxy-α-[(methylamino)methyl]-, hydrochloride (9CI),Benzyl alcohol, p-hydroxy-α-[(methylamino)methyl]-, hydrochloride (8CI),Oxedrine hydrochloride,Synephrine hydrochloride,p-Hydroxy-α-(methylaminomethyl)benzyl alcohol hydrochloride IUPAC Name: 4-[1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride SMILES: Cl.CNCC(O)c1ccc(O)cc1