accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 59865-82-6

SSNH2

CAS RN 59865-82-6

IUPAC Name: 2-(phenylsulfanyl)ethanethioamide
Synonyms: 2-(phenylsulfanyl)ethanethioamide
Molecular Weight (g/mol): 183.29
Molecular Formula: C8H9NS2
InChi Key: ZRDOSZLCJHRADP-UHFFFAOYSA-N
SMILES: NC(=S)CSC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results