accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 59865-87-1

IUPAC Name: 2-(4-chlorobenzenesulfonyl)ethanethioamide
Synonyms: 2-(4-chlorobenzenesulfonyl)ethanethioamide
Molecular Weight (g/mol): 249.73
Molecular Formula: C8H8ClNO2S2
InChi Key: OGQJCNYNVJXWBB-UHFFFAOYSA-N
SMILES: NC(=S)CS(=O)(=O)C1=CC=C(Cl)C=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results