accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5988-19-2

IUPAC Name: (4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid
Synonyms: L-dihydroorotic acid
Molecular Weight (g/mol): 158.11
Molecular Formula: C5H6N2O4
InChi Key: UFIVEPVSAGBUSI-REOHCLBHSA-N
SMILES: OC(=O)[C@@H]1CC(=O)NC(=O)N1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results