accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 599-04-2

IUPAC Name: 3-hydroxy-4,4-dimethyloxolan-2-one
Synonyms: pantolactone
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: SERHXTVXHNVDKA-UHFFFAOYNA-N
SMILES: CC1(C)COC(=O)C1O
Molecular Weight (g/mol) 
  • (7)
  • (2)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (7)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)