accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 599-91-7

IUPAC Name: propyl 4-methylbenzene-1-sulfonate
Synonyms: propyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 214.28
Molecular Formula: C10H14O3S
InChi Key: JTTWNTXHFYNETH-UHFFFAOYSA-N
SMILES: CCCOS(=O)(=O)C1=CC=C(C)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results