accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 599-94-0

IUPAC Name: [2-(benzenesulfonyl)ethanesulfonyl]benzene
Synonyms: [2-(benzenesulfonyl)ethanesulfonyl]benzene
Molecular Weight (g/mol): 310.38
Molecular Formula: C14H14O4S2
InChi Key: ULELOBVZIKJPAC-UHFFFAOYSA-N
SMILES: O=S(=O)(CCS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results