accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 599191-73-8

IUPAC Name: 4-iodo-1H-indazol-3-amine
Synonyms: 4-iodo-1H-indazol-3-amine
Molecular Weight (g/mol): 259.05
Molecular Formula: C7H6IN3
InChi Key: OEQIVIYSUJXCFG-UHFFFAOYSA-N
SMILES: NC1=NNC2=CC=CC(I)=C12
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results