accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 59936-29-7

IUPAC Name: 1-tert-butyl 2-methyl (2S)-pyrrolidine-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-methyl (2S)-pyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 229.28
Molecular Formula: C11H19NO4
InChi Key: WVDGSSCWFMSRHN-QMMMGPOBSA-N
SMILES: COC(=O)[C@@H]1CCCN1C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results