accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNSH2N

CAS RN 59944-75-1

IUPAC Name: thieno[2,3-b]pyrazin-7-amine
Synonyms: thieno[2,3-b]pyrazin-7-amine
Molecular Weight (g/mol): 151.19
Molecular Formula: C6H5N3S
InChi Key: ZRVKSPNBHZCQKY-UHFFFAOYSA-N
SMILES: NC1=CSC2=NC=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results