accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5995-86-8

IUPAC Name: 3,4,5-trihydroxybenzoate
Synonyms: gallate
Molecular Weight (g/mol): 169.11
Molecular Formula: C7H5O5
InChi Key: LNTHITQWFMADLM-UHFFFAOYSA-M
SMILES: OC1=CC(=CC(O)=C1O)C([O-])=O
Molecular Weight (g/mol) 
  • (10)
  • (1)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (2)
Grade 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (1)
  • (10)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (4)
  • (1)