accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOOCH3OOCH3

CAS RN 60-01-5

IUPAC Name: 1,3-bis(butanoyloxy)propan-2-yl butanoate
Synonyms: tributyrin glyceroltributyrin butyric acid triester with glycerin tri-butyrin glyceryl tributyrate butyrin glycerin tributyrate butyrin, tri- tributyrinine tributyrl glyceride
Molecular Weight (g/mol): 302.37
Molecular Formula: C15H26O6
InChi Key: UYXTWWCETRIEDR-UHFFFAOYSA-N
SMILES: CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (6)
  • (6)