accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NONH2N

CAS RN 60-27-5

IUPAC Name: 2-amino-1-methyl-4,5-dihydro-1H-imidazol-4-one
Synonyms: 1-methylhydantoin-2-imide
Molecular Weight (g/mol): 113.12
Molecular Formula: C4H7N3O
InChi Key: DDRJAANPRJIHGJ-UHFFFAOYSA-N
SMILES: CN1CC(=O)N=C1N
Quantity 
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (11)
Percent Purity 
  • (3)
  • (8)
  • (3)
Physical Form 
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (11)
  • (3)
  • (8)
  • (3)
  • (3)
  • (3)
  • (3)