accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NOHO

CAS RN 60-32-2

IUPAC Name: 6-aminohexanoic acid
Synonyms: aminocaproic acid epsilon-amino-n-hexanoic acid epsilon-aminocapronzuur afibrin aminocaproic acid (inn) and its salts epsilcapramin 氨基己酸 caprolisin hemocaprol H-epsilon-acp-OH
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: SLXKOJJOQWFEFD-UHFFFAOYSA-N
SMILES: NCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (8)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (7)
  • (1)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (4)
  • (3)
  • (3)
  • (3)
  • (8)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (7)
  • (1)
  • (3)
  • (4)