accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 60-51-5

IUPAC Name: O,O-dimethyl {[(methylcarbamoyl)methyl]sulfanyl}phosphonothioate
Synonyms: dimethoate phosphamide
Molecular Weight (g/mol): 229.25
Molecular Formula: C5H12NO3PS2
InChi Key: MCWXGJITAZMZEV-UHFFFAOYSA-N
SMILES: CNC(=O)CSP(=S)(OC)OC
Molecular Weight (g/mol) 
  • (4)
  • (5)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Concentration 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)