accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNOH3C

CAS RN 60-80-0

IUPAC Name: 1,5-dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
Synonyms: antipyrine phenazone (pharmaceutical) 安替比林 antipyrinum anodynine antipyrine (substance) acetasol azophen hloralantipyrine antipyrine (product)
Molecular Weight (g/mol): 188.23
Molecular Formula: C11H12N2O
InChi Key: VEQOALNAAJBPNY-UHFFFAOYSA-N
SMILES: CN1N(C(=O)C=C1C)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)