accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6000-43-7

IUPAC Name: hydrogen 2-aminoacetic acid chloride
Synonyms: glycine hcl
Molecular Weight (g/mol): 111.53
Molecular Formula: C2H6ClNO2
InChi Key: IVLXQGJVBGMLRR-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC(O)=O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)