accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNO

CAS RN 6002-15-9

IUPAC Name: 1-phenyl-1H-imidazole-2-carbaldehyde
Synonyms: 1-phenylimidazole-2-carbaldehyde
Molecular Weight (g/mol): 172.19
Molecular Formula: C10H8N2O
InChi Key: VJDRBJPZIHUDNE-UHFFFAOYSA-N
SMILES: O=CC1=NC=CN1C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results