accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6003-94-7

OOOOOO

CAS RN 6003-94-7

IUPAC Name: 4-oxo-4H-pyran-2,6-dicarboxylate
Synonyms: 4-oxopyran-2,6-dicarboxylate
Molecular Weight (g/mol): 182.09
Molecular Formula: C7H2O6
InChi Key: PBAYDYUZOSNJGU-UHFFFAOYSA-L
SMILES: [O-]C(=O)C1=CC(=O)C=C(O1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results